accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClClO

CAS RN 6361-23-5

IUPAC Name: 2,5-dichlorobenzaldehyde
Synonyms: 2,5-dichloro-benzaldehyde
Molecular Weight (g/mol): 175.01
Molecular Formula: C7H4Cl2O
InChi Key: BUXHYMZMVMNDMG-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C(C=O)=C1
Boiling Point 
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)