accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 636-73-7

OSOON

CAS RN 636-73-7

IUPAC Name: pyridine-3-sulfonate
Synonyms: pyridine-3-sulfonate
Molecular Weight (g/mol): 158.15
Molecular Formula: C5H4NO3S
InChi Key: DVECLMOWYVDJRM-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C1=CC=CN=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)