accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 635-90-5

IUPAC Name: 1-phenyl-1H-pyrrole
Synonyms: pyrrole, 1-phenyl-
Molecular Weight (g/mol): 143.19
Molecular Formula: C10H9N
InChi Key: GEZGAZKEOUKLBR-UHFFFAOYSA-N
SMILES: C1=CN(C=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results