accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 635-39-2

NH2NOHNNHNHCl

CAS RN 635-39-2

IUPAC Name: hydrogen 2-amino-6,7-dihydro-3H-purin-6-one chloride
Synonyms: hydrogen 2-aminohypoxanthine chloride
Molecular Weight (g/mol): 187.59
Molecular Formula: C5H6ClN5O
InChi Key: IBAOFQIOOBQLHE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=NC(=O)C2=C(N1)N=CN2
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results