accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 635-12-1

OO

CAS RN 635-12-1

IUPAC Name: 1,4-dihydroanthracene-1,4-dione
Synonyms: anthracene-1,4-dione
Molecular Weight (g/mol): 208.22
Molecular Formula: C14H8O2
InChi Key: LSOTZYUVGZKSHR-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)C2=C1C=C1C=CC=CC1=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results