accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHN

CAS RN 6343-54-0

IUPAC Name: N-benzylformamide
Synonyms: N-benzylformamide
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: IIBOGKHTXBPGEI-UHFFFAOYSA-N
SMILES: O=CNCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)