accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 63370-90-1

H3CCH3H3COHCOCH3CH3H3CH3CCH3H3COHCOCH3CH3H3CH3CCH3H3COHCOCH3CH3H3CH3CCH3H3COHCOCH3CH3H3CHf

CAS RN 63370-90-1

IUPAC Name: hafnium(4+) tetrakis(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Synonyms: hafnium(4+) tetrakis(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Molecular Weight (g/mol): 911.57
Molecular Formula: C44H76HfO8
InChi Key: GPCQYWUYUFMJFR-UHFFFAOYSA-N
SMILES: [Hf+4].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results