accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NHN

CAS RN 63361-56-8

IUPAC Name: benzyl[(pyridin-3-yl)methyl]amine
Synonyms: benzyl(pyridin-3-ylmethyl)amine
Molecular Weight (g/mol): 198.27
Molecular Formula: C13H14N2
InChi Key: COHYOBKZKMKMIX-UHFFFAOYSA-N
SMILES: C(NCC1=CC=CN=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results