accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 63211-98-3

IUPAC Name: 2-chloro-6-methylpyrimidine-4,5-diamine
Synonyms: 2-chloro-6-methylpyrimidine-4,5-diamine
Molecular Weight (g/mol): 158.59
Molecular Formula: C5H7ClN4
InChi Key: KSGKEVCHVYUUDN-UHFFFAOYSA-N
SMILES: CC1=NC(Cl)=NC(N)=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results