accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6314-42-7

H2NOS

CAS RN 6314-42-7

IUPAC Name: 1-benzothiophene-2-carboxamide
Synonyms: 1-benzothiophene-2-carboxamide
Molecular Weight (g/mol): 177.22
Molecular Formula: C9H7NOS
InChi Key: GYSCBCSGKXNZRH-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC2=CC=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results