accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OFFOHO(E/Z)

CAS RN 630424-87-2

IUPAC Name: 3-[3-(difluoromethoxy)phenyl]prop-2-enoic acid
Synonyms: 3-[3-(difluoromethoxy)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 214.17
Molecular Formula: C10H8F2O3
InChi Key: YAXXERXTVQNNBQ-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CC(OC(F)F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results