accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 630117-19-0

CH3OHNNNNOOHOOH(E/Z)

CAS RN 630117-19-0

IUPAC Name: N-{[4-(2-cyanophenyl)piperazin-1-yl]methyl}-3-methylbenzamide; but-2-enedioic acid
Synonyms: N-{[4-(2-cyanophenyl)piperazin-1-yl]methyl}-3-methylbenzamide; butenedioic acid
Molecular Weight (g/mol): 450.5
Molecular Formula: C24H26N4O5
InChi Key: NAEUGRPISCANHO-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(O)=O.CC1=CC=CC(=C1)C(=O)NCN1CCN(CC1)C1=CC=CC=C1C#N
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results