accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 63-84-3

NH2HOOHOHO

CAS RN 63-84-3

IUPAC Name: 2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
Synonyms: 3',4'-dihydroxyphenylalanine dopastone helfo dopa 3-hydroxytyrosine cerepap dopicar dopaston SE 3-hydroxy-DL-tyrosine L-(o-dihydroxyphenyl)alanine L-O-dihydroxyphenylalanine
Molecular Weight (g/mol): 197.19
Molecular Formula: C9H11NO4
InChi Key: WTDRDQBEARUVNC-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(O)C(O)=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)