accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6299-85-0

CH3OOClNClN

CAS RN 6299-85-0

IUPAC Name: methyl 2,6-dichloropyrimidine-4-carboxylate
Synonyms: methyl 2,6-dichloropyrimidine-4-carboxylate
Molecular Weight (g/mol): 207.01
Molecular Formula: C6H4Cl2N2O2
InChi Key: DQNQNLWKAGZNIT-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=NC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results