accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHN

CAS RN 6293-56-7

IUPAC Name: 2-(pyridin-3-yl)ethan-1-ol
Synonyms: 3-pyridineethanol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: YPWSASPSYAWQRK-UHFFFAOYSA-N
SMILES: OCCC1=CC=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results