accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH

CAS RN 629-89-0

IUPAC Name: octadec-1-yne
Synonyms: octadec-1-yne
Molecular Weight (g/mol): 250.47
Molecular Formula: C18H34
InChi Key: IYDNQWWOZQLMRH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)