accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrOOCl

CAS RN 62878-96-0

IUPAC Name: 5-bromo-1-benzofuran-2-carbonyl chloride
Synonyms: 5-bromo-1-benzofuran-2-carbonyl chloride
Molecular Weight (g/mol): 259.48
Molecular Formula: C9H4BrClO2
InChi Key: FGPAEHRRVKLUHA-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC2=CC(Br)=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results