accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNH2NCH3

CAS RN 62821-88-9

IUPAC Name: 2-(3,5-dimethyl-1H-pyrazol-1-yl)ethan-1-amine
Synonyms: 2-(3,5-dimethylpyrazol-1-yl)ethanamine
Molecular Weight (g/mol): 139.2
Molecular Formula: C7H13N3
InChi Key: FXLOXQBWUPCCDL-UHFFFAOYSA-N
SMILES: CC1=NN(CCN)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results