accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62813-01-8

IUPAC Name: 1-cyclopropylpiperidin-4-one
Synonyms: 1-cyclopropylpiperidin-4-one
Molecular Weight (g/mol): 139.2
Molecular Formula: C8H13NO
InChi Key: DTUJRJIWGWTNFQ-UHFFFAOYSA-N
SMILES: O=C1CCN(CC1)C1CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results