accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COClHOO

CAS RN 6280-89-3

IUPAC Name: 2-chloro-5-methoxybenzoic acid
Synonyms: 2-chloro-5-methoxybenzoic acid
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: AQHFCRYZABKUEV-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results