accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 627871-16-3

CH3H2NBrF

CAS RN 627871-16-3

IUPAC Name: 5-bromo-4-fluoro-2-methylaniline
Synonyms: 5-bromo-4-fluoro-2-methylaniline
Molecular Weight (g/mol): 204.04
Molecular Formula: C7H7BrFN
InChi Key: DNCLVDGUXUSPTL-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(Br)C=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results