accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 627-09-8

H3COOCH

CAS RN 627-09-8

IUPAC Name: prop-2-yn-1-yl acetate
Synonyms: 2-propyn-1-ol, acetate
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: RIZZXCJMFIGMON-UHFFFAOYSA-N
SMILES: CC(=O)OCC#C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)