accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NHCH3

CAS RN 626-13-1

IUPAC Name: 3-methyl-N-(3-methylphenyl)aniline
Synonyms: 3-methyl-N-(3-methylphenyl)aniline
Molecular Weight (g/mol): 197.28
Molecular Formula: C14H15N
InChi Key: CWVPIIWMONJVGG-UHFFFAOYSA-N
SMILES: CC1=CC(NC2=CC(C)=CC=C2)=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results