accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2ClClFFF

CAS RN 62593-17-3

IUPAC Name: 2,4-dichloro-6-(trifluoromethyl)aniline
Synonyms: 2,4-dichloro-6-(trifluoromethyl)aniline
Molecular Weight (g/mol): 230.01
Molecular Formula: C7H4Cl2F3N
InChi Key: OCJPQZFOBGQYEA-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(Cl)C=C1C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results