accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6258-66-8

HSCl

CAS RN 6258-66-8

IUPAC Name: (4-chlorophenyl)methanethiol
Synonyms: 4-chlor-benzylthiol
Molecular Weight (g/mol): 158.64
Molecular Formula: C7H7ClS
InChi Key: GKQXPTHQTXCXEV-UHFFFAOYSA-N
SMILES: SCC1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)