accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62564-90-3

CH3NNONH2

CAS RN 62564-90-3

IUPAC Name: 5-amino-1-methyl-1H-pyrazole-4-carbaldehyde
Synonyms: 5-amino-1-methylpyrazole-4-carbaldehyde
Molecular Weight (g/mol): 125.13
Molecular Formula: C5H7N3O
InChi Key: NDGIYFSWJGZTRC-UHFFFAOYSA-N
SMILES: CN1N=CC(C=O)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results