accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 625-95-6

IUPAC Name: 1-iodo-3-methylbenzene
Synonyms: benzene, 1-iodo-3-methyl-
Molecular Weight (g/mol): 218.04
Molecular Formula: C7H7I
InChi Key: VLCPISYURGTGLP-UHFFFAOYSA-N
SMILES: CC1=CC=CC(I)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results