accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 625-80-9

IUPAC Name: 2-(propan-2-ylsulfanyl)propane
Synonyms: 2,4-dimethyl-3-thiapentane
Molecular Weight (g/mol): 118.24
Molecular Formula: C6H14S
InChi Key: XYWDPYKBIRQXQS-UHFFFAOYSA-N
SMILES: CC(C)SC(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results