accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 625-38-7

OHOH2C

CAS RN 625-38-7

IUPAC Name: but-3-enoic acid
Synonyms: 3-butenoic acid vinyl acetic acid allylic acid vinylessigsaeure
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: PVEOYINWKBTPIZ-UHFFFAOYSA-N
SMILES: OC(=O)CC=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)