accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 625-08-1

H3CH3COHOHO

CAS RN 625-08-1

IUPAC Name: 3-hydroxy-3-methylbutanoic acid
Synonyms: 3-hydroxyisovaleric acid 3-hydroxy-isovaleric acid HMB hmb-d6 3-OH-isovaleric acid β-hydroxyisovaleric acid
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: AXFYFNCPONWUHW-UHFFFAOYSA-N
SMILES: CC(C)(O)CC(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (4)
  • (1)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (5)
  • (4)
  • (1)
  • (1)