accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62482-30-8

H3COOH3CNBr

CAS RN 62482-30-8

IUPAC Name: ethyl 6-bromo-2-methylquinoline-4-carboxylate
Synonyms: ethyl 6-bromo-2-methylquinoline-4-carboxylate
Molecular Weight (g/mol): 294.15
Molecular Formula: C13H12BrNO2
InChi Key: KOVIYCANWGLBHA-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C2C=C(Br)C=CC2=NC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results