accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COO

CAS RN 6243-10-3

IUPAC Name: cyclohexyl hexanoate
Synonyms: cyclohexyl hexanoate
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: DTNOERNOMHQUCN-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OC1CCCCC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results