accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 624-28-2

BrBrN

CAS RN 624-28-2

IUPAC Name: 2,5-dibromopyridine
Synonyms: 2,5-dibromopyridine
Molecular Weight (g/mol): 236.89
Molecular Formula: C5H3Br2N
InChi Key: ZHXUWDPHUQHFOV-UHFFFAOYSA-N
SMILES: BrC1=CC=C(Br)N=C1
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)