accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6234-01-1

OO(S)OONH2H3CH3CCH3H3CHCl

CAS RN 6234-01-1

IUPAC Name: 5-tert-butyl 1-methyl (2S)-2-aminopentanedioate hydrochloride
Synonyms: 5-tert-butyl 1-methyl (2S)-2-aminopentanedioate hydrochloride
Molecular Weight (g/mol): 253.72
Molecular Formula: C10H20ClNO4
InChi Key: YIFPACFSZQWAQF-RZUXQYDTNA-N
SMILES: Cl.COC(=O)[C@@H](N)CCC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)