accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 623-98-3

H3COSOOCH3

CAS RN 623-98-3

IUPAC Name: dipropyl sulfite
Synonyms: dipropyl sulfite
Molecular Weight (g/mol): 166.24
Molecular Formula: C6H14O3S
InChi Key: MAIQPVFXODAAIG-UHFFFAOYSA-N
SMILES: CCCOS(=O)OCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)