accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 623-17-6

H3COOO

CAS RN 623-17-6

IUPAC Name: (furan-2-yl)methyl acetate
Synonyms: 2-furanmethanol, acetate
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: CKOYRRWBOKMNRG-UHFFFAOYSA-N
SMILES: CC(=O)OCC1=CC=CO1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results