accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62284-79-1

HOOOHOBrNSN

CAS RN 62284-79-1

IUPAC Name: 6-(4-bromophenyl)-2H,3H,5H,6H-imidazo[2,1-b][1,3]thiazole; oxalic acid
Synonyms: 6-(4-bromophenyl)-2H,3H,5H,6H-imidazo[2,1-b][1,3]thiazole; oxalic acid
Molecular Weight (g/mol): 373.22
Molecular Formula: C13H13BrN2O4S
InChi Key: ZULBIBHDIQCNIS-UHFFFAOYNA-N
SMILES: OC(=O)C(O)=O.BrC1=CC=C(C=C1)C1CN2CCSC2=N1
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results