accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62235-61-4

BrNNHO

CAS RN 62235-61-4

IUPAC Name: 3-(bromomethyl)-1,2-dihydroquinoxalin-2-one
Synonyms: 3-(bromomethyl)-1H-quinoxalin-2-one
Molecular Weight (g/mol): 239.07
Molecular Formula: C9H7BrN2O
InChi Key: SOTAKCFOTFCVSS-UHFFFAOYSA-N
SMILES: BrCC1=NC2=CC=CC=C2NC1=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results