accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62202-86-2

H3CMgCH3

CAS RN 62202-86-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 110.48
Molecular Formula: C6H14Mg
InChi Key: YHNWUQFTJNJVNU-UHFFFAOYSA-N
SMILES: [Mg++].C[CH2-].CCC[CH2-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results