accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 622-75-3

NN

CAS RN 622-75-3

IUPAC Name: 2-[4-(cyanomethyl)phenyl]acetonitrile
Synonyms: 2-[4-(cyanomethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 156.19
Molecular Formula: C10H8N2
InChi Key: FUQCKESKNZBNOG-UHFFFAOYSA-N
SMILES: N#CCC1=CC=C(CC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results