accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 621-33-0

CH3OH2N

CAS RN 621-33-0

IUPAC Name: 3-ethoxyaniline
Synonyms: 3-ethoxyaniline
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: WEZAHYDFZNTGKE-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results