accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 621-29-4

IUPAC Name: 1-isocyanato-3-methylbenzene
Synonyms: 1-isocyanato-3-methylbenzene
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: CPPGZWWUPFWALU-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results