accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HO

CAS RN 621-27-2

IUPAC Name: 3-propylphenol
Synonyms: 3-propylphenol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: MPWGZBWDLMDIHO-UHFFFAOYSA-N
SMILES: CCCC1=CC=CC(O)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results