accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6209-44-5

ONHOHNONOOH2OH2OH2O

CAS RN 6209-44-5

IUPAC Name: 5-nitro-1,3-diazinane-2,4,6-trione trihydrate
Synonyms: 5-nitro-1,3-diazinane-2,4,6-trione trihydrate
Molecular Weight (g/mol): 227.13
Molecular Formula: C4H9N3O8
InChi Key: MXFRQEURRMBYRS-UHFFFAOYSA-N
SMILES: O.O.O.[O-][N+](=O)C1C(=O)NC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results