accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NClFFF

CAS RN 62039-92-3

IUPAC Name: 1-[4-chloro-3-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[4-chloro-3-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 209.6
Molecular Formula: C8H7ClF3N
InChi Key: SRHQOQMNBVOXCF-UHFFFAOYSA-N
SMILES: NCC1=CC=C(Cl)C(=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results