accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 620-84-8

CH3NH

CAS RN 620-84-8

IUPAC Name: 4-methyl-N-phenylaniline
Synonyms: 4-methyl-N-phenylbenzamine
Molecular Weight (g/mol): 183.25
Molecular Formula: C13H13N
InChi Key: AGHYMXKKEXDUTA-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results