accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 620-14-4

H3CH3C

CAS RN 620-14-4

IUPAC Name: 1-ethyl-3-methylbenzene
Synonyms: M-ethyltoluene
Molecular Weight (g/mol): 120.2
Molecular Formula: C9H12
InChi Key: ZLCSFXXPPANWQY-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(C)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)