accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONBr

CAS RN 61995-52-6

IUPAC Name: 1-(5-bromo-1H-indol-1-yl)ethan-1-one
Synonyms: 1-(5-bromoindol-1-yl)ethanone
Molecular Weight (g/mol): 238.08
Molecular Formula: C10H8BrNO
InChi Key: BOMKWHSZGCMFEG-UHFFFAOYSA-N
SMILES: CC(=O)N1C=CC2=CC(Br)=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results