CAS RN 61948-85-4
CAS RN 61948-85-4
IUPAC Name:
2-amino-3,4,5-trimethoxybenzoic acid
Synonyms:
2-amino-3,4,5-trimethoxybenzoic acid
Molecular Weight (g/mol):
227.22
Molecular Formula:
C10H13NO5
InChi Key:
JSHSRQCOCMIIPA-UHFFFAOYSA-N
SMILES:
COC1=CC(C(O)=O)=C(N)C(OC)=C1OC
1
–
1
of
1
results
1
2-Amino-3,4,5-trimethoxybenzoic acid, 97%
CAS: 61948-85-4 Molecular Formula: C10H13NO5 MDL Number: MFCD00051630 InChI Key: JSHSRQCOCMIIPA-UHFFFAOYSA-N PubChem CID: 96602 IUPAC Name: 2-amino-3,4,5-trimethoxybenzoic acid