accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 619-57-8

H2NOOH

CAS RN 619-57-8

IUPAC Name: 4-hydroxybenzamide
Synonyms: 4-hydroxybenzamide P-hydroxybenzamide
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: QXSAKPUBHTZHKW-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results